Assay Details
Target Gene Details
Entrez Gene ID: | 83706 |
Gene Name: | fermitin family member 3 |
Gene Aliases: |
KIND3, MIG-2, MIG2B, UNC112C, URP2, URP2SF |
Location: |
Chr.11:64206665-64223891 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FERMT3 | NM_031471.5 | 8 | 1130 | NP_113659.3 |
NM_178443.2 | 8 | 1130 | NP_848537.1 | |
XM_011545294.2 | 8 | 1142 | XP_011543596.1 | |
XM_011545295.2 | 7 | 975 | XP_011543597.1 | |
XM_017018398.1 | 8 | 1138 | XP_016873887.1 | |
XM_017018399.1 | 7 | 990 | XP_016873888.1 | |
AK093719.1 | 8 | 1075 | BAC04220.1 | |
AY093951.1 | 8 | 1076 | AAM19736.1 | |
AY093952.1 | 8 | 1076 | AAM19737.1 | |
BC004347.1 | 8 | 1074 | AAH04347.2 | |
BC013366.2 | 8 | 1074 | AAH13366.1 | |
BC015584.2 | 8 | 1069 | AAH15584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1159904 | Chr.11:64150129 - 64279169 on Build GRCh38 | Deletion | PPP1R14B DNAJC4 TRPT1 GPR137 STIP1 MACROD1 NUDT22 BAD LOC105369340 FERMT3 PLCB3 FKBP2 VEGFB |
nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion | MIR7155 NUDT22 ESRRA PRDX5 BAD TEX40 FERMT3 PLCB3 FKBP2 KCNK4 FLRT1 VEGFB PPP1R14B DNAJC4 TRPT1 GPR137 STIP1 COX8A OTUB1 MACROD1 CCDC88B TRMT112 LOC105369340 NAA40 KCNK4-TEX40 |
dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss | PPP1R14B DNAJC4 TRPT1 GPR137 STIP1 NUDT22 ESRRA PRDX5 BAD TRMT112 TEX40 LOC105369340 FERMT3 PLCB3 FKBP2 KCNK4 KCNK4-TEX40 VEGFB |
nsv555187 | Chr.11:64185649 - 64260416 on Build GRCh38 | Loss | PPP1R14B DNAJC4 TRPT1 LOC105369340 FERMT3 STIP1 PLCB3 FKBP2 NUDT22 VEGFB |
nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss | PPP1R14B DNAJC4 TRPT1 GPR137 MIR7155 NUDT22 CCDC88B ESRRA PRDX5 BAD TRMT112 MIR1237 TEX40 LOC105369340 FERMT3 PLCB3 FKBP2 KCNK4 RPS6KA4 KCNK4-TEX40 VEGFB |
nsv555184 | Chr.11:64156974 - 64275525 on Build GRCh38 | Loss | PPP1R14B DNAJC4 TRPT1 GPR137 STIP1 MACROD1 NUDT22 BAD LOC105369340 FERMT3 PLCB3 FKBP2 VEGFB |
nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss | PPP1R14B DNAJC4 TRPT1 GPR137 STIP1 MACROD1 NUDT22 ESRRA PRDX5 BAD TRMT112 TEX40 LOC105369340 FERMT3 PLCB3 FKBP2 KCNK4 FLRT1 KCNK4-TEX40 VEGFB |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |