Assay Details
Target Gene Details
Entrez Gene ID: | 10140 |
Gene Name: | transducer of ERBB2, 1 |
Gene Aliases: |
APRO5, APRO6, PIG49, TOB, TROB, TROB1 |
Location: |
Chr.17:50862223-50867978 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB1 | NM_001243877.1 | 3 | 1438 | NP_001230806.1 |
NM_001243885.1 | 2 | 1249 | NP_001230814.1 | |
NM_005749.3 | 2 | 1452 | NP_005740.1 | |
AK313904.1 | ||||
AY524047.1 | 1 | 1019 | AAS94256.1 | |
BC031406.1 | 1 | 1016 | AAH31406.1 | |
BC070493.1 | AAH70493.1 | |||
BC098415.1 | AAH98415.1 | |||
BT019380.1 | AAV38187.1 | |||
BT019381.1 | AAV38188.1 | |||
CR536487.1 | CAG38726.1 | |||
CR541686.1 | CAG46487.1 | |||
D38305.1 | 1 | 1053 | BAA07423.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |