Assay Details
Target Gene Details
Entrez Gene ID: | 57611 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat 2 |
Gene Aliases: |
LINX |
Location: |
Chr.15:74128203-74141826 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ISLR2 | NM_001130136.1 | 4 | 3895 | NP_001123608.1 |
NM_001130137.1 | 4 | 3631 | NP_001123609.1 | |
NM_001130138.1 | 4 | 3339 | NP_001123610.1 | |
NM_020851.2 | 3 | 3464 | NP_065902.1 | |
XM_011521840.2 | 4 | 3734 | XP_011520142.1 | |
XM_011521841.1 | 4 | 3417 | XP_011520143.1 | |
XM_017022446.1 | XP_016877935.1 | |||
AB040898.2 | 4 | 3895 | ||
AK092297.1 | ||||
AK092302.1 | ||||
AK096602.1 | ||||
AY358316.1 | 3 | 3262 | AAQ88682.1 | |
BC114551.1 | 3 | 1978 | AAI14552.1 | |
BC152429.1 | 4 | 3895 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv827385 | Chr.15:74127198 - 74136960 on Build GRCh38 | Gain |
![]() ![]() |
nsv951993 | Chr.15:74124460 - 74137359 on Build GRCh38 | Deletion |
![]() ![]() |
esv2760040 | Chr.15:73922773 - 74172575 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1047564 | Chr.15:74116495 - 74165840 on Build GRCh38 | Gain |
![]() ![]() |
nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)