Assay Details
Target Gene Details
Entrez Gene ID: | 13 |
Gene Name: | arylacetamide deacetylase |
Gene Aliases: |
CES5A1, DAC |
Location: |
Chr.3:151814008-151828488 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AADAC | NM_001086.2 | 4 | 656 | NP_001077.2 |
XM_005247104.4 | 4 | 670 | XP_005247161.1 | |
AK290628.1 | 4 | 607 | ||
AV705031.1 | 4 | 656 | ||
BC032309.1 | 4 | 591 | AAH32309.1 | |
L32179.1 | 4 | 651 | AAA35551.1 |
Target Gene Details
Entrez Gene ID: | 101928142 |
Gene Name: | AADACL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:151751179-151928175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AADACL2-AS1 | NR_110202.1 | |||
NR_110203.1 | ||||
DB454604.1 | ||||
HY022483.1 | ||||
HY205836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv622n27 | Chr.3:151796802 - 151877081 on Build GRCh38 | Loss | AADACL2-AS1 SUCNR1 AADAC |
nsv592070 | Chr.3:151817634 - 151828253 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
nsv592072 | Chr.3:151821400 - 151828253 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
nsv522472 | Chr.3:151743597 - 151864407 on Build GRCh38 | Loss | AADACL2-AS1 AADACL2 AADAC AADACP1 |
dgv1419e212 | Chr.3:151792302 - 151831226 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
esv2759189 | Chr.3:151757492 - 151945387 on Build GRCh38 | Loss | AADACL2-AS1 SUCNR1 AADACL2 AADAC AADACP1 |
esv25716 | Chr.3:151793701 - 151833212 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
esv2763771 | Chr.3:151790048 - 151837943 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
dgv8655n54 | Chr.3:151814106 - 151833114 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
dgv4920n100 | Chr.3:151784289 - 151830691 on Build GRCh38 | Loss | AADACL2-AS1 AADAC AADACP1 |
esv3562727 | Chr.3:151792826 - 151836712 on Build GRCh38 | Deletion | AADACL2-AS1 AADAC |
esv2672544 | Chr.3:151793742 - 151833274 on Build GRCh38 | Deletion | AADACL2-AS1 AADAC |
nsv514175 | Chr.3:151793730 - 151832482 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
esv3598188 | Chr.3:151793743 - 151833279 on Build GRCh38 | Loss | AADACL2-AS1 AADAC |
dgv8654n54 | Chr.3:151792469 - 151877081 on Build GRCh38 | Loss | AADACL2-AS1 SUCNR1 AADAC |
dgv4919n100 | Chr.3:151776103 - 151841876 on Build GRCh38 | Loss | AADACL2-AS1 AADAC AADACP1 |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |