Assay Details
Target Gene Details
Entrez Gene ID: | 7384 |
Gene Name: | ubiquinol-cytochrome c reductase core protein I |
Gene Aliases: |
D3S3191, QCR1, UQCR1 |
Location: |
Chr.3:48598999-48609665 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UQCRC1 | NM_003365.2 | 8 | 952 | NP_003356.2 |
AK129903.1 | 8 | 932 | ||
AK300694.1 | 8 | 887 | ||
AK313090.1 | 8 | 922 | ||
BC009586.1 | 8 | 923 | AAH09586.1 | |
D26485.1 | 8 | 914 | BAA05495.1 | |
L16842.1 | 8 | 1324 | AAA20046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954484 | Chr.3:48533268 - 48609567 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1004383 | Chr.3:48506361 - 48612339 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv3811 | Chr.3:48561747 - 48606376 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv834685 | Chr.3:48527576 - 48708341 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv818138 | Chr.3:48573492 - 48694054 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv590246 | Chr.3:48590581 - 48683607 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)