Assay Details
Target Gene Details
Entrez Gene ID: | 860 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA |
Location: |
Chr.6:45328142-45664032 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RUNX2 | NM_001015051.3 | NP_001015051.3 | ||
NM_001024630.3 | NP_001019801.3 | |||
NR_103532.1 | ||||
NR_103533.1 | ||||
XM_011514960.2 | XP_011513262.1 | |||
XM_011514961.2 | XP_011513263.1 | |||
XM_011514962.2 | XP_011513264.1 | |||
XM_011514963.2 | XP_011513265.1 | |||
XM_011514964.2 | XP_011513266.1 | |||
XM_011514965.2 | XP_011513267.1 | |||
XM_017011391.1 | XP_016866880.1 | |||
XM_017011393.1 | XP_016866882.1 | |||
XM_017011394.1 | XP_016866883.1 | |||
XM_017011395.1 | XP_016866884.1 | |||
AW469546.1 | ||||
BX108677.1 |
Target Gene Details
Entrez Gene ID: | 8464 |
Gene Name: | SPT3 homolog, SAGA and STAGA complex component |
Gene Aliases: |
SPT3, SPT3L |
Location: |
Chr.6:44808844-45378051 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SUPT3H | NM_001261823.1 | NP_001248752.1 | ||
NM_003599.3 | NP_003590.1 | |||
NM_181356.2 | NP_852001.1 | |||
XM_011514949.2 | XP_011513251.1 | |||
XM_011514950.1 | XP_011513252.1 | |||
XM_011514951.2 | XP_011513253.1 | |||
XM_011514952.2 | XP_011513254.1 | |||
XM_011514953.2 | XP_011513255.1 | |||
XM_011514954.2 | XP_011513256.1 | |||
XM_017011369.1 | XP_016866858.1 | |||
XM_017011371.1 | XP_016866860.1 | |||
XM_017011372.1 | XP_016866861.1 | |||
XM_017011373.1 | XP_016866862.1 | |||
AF069734.1 | AAC39904.1 | |||
AF073930.1 | AAC36098.1 | |||
AK303838.1 | ||||
AK313874.1 | ||||
BC035816.1 | AAH35816.1 | |||
BC050384.1 | AAH50384.1 | |||
DA012115.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |