Assay Details
Target Gene Details
Entrez Gene ID: | 123811 |
Gene Name: | FGFR1OP N-terminal like |
Gene Aliases: |
C16orf63, FOR20, PHSECRG2 |
Location: |
Chr.16:15865719-15888649 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FOPNL | NM_001304497.1 | 2 | 169 | NP_001291426.1 |
NM_001304498.1 | 2 | 169 | NP_001291427.1 | |
NM_001304499.1 | 2 | 169 | NP_001291428.1 | |
NM_001304500.1 | NP_001291429.1 | |||
NM_001304502.1 | 2 | 169 | NP_001291431.1 | |
NM_144600.3 | 2 | 169 | NP_653201.1 | |
NR_130755.1 | 2 | 169 | ||
NR_130756.1 | 2 | 169 | ||
NR_130757.1 | ||||
AK055715.1 | 2 | 112 | BAB70994.1 | |
AK056798.1 | 2 | 122 | ||
AL832498.1 | 2 | 132 | ||
AY507846.1 | 2 | 125 | AAR98812.1 | |
BC022321.1 | 2 | 120 | AAH22321.1 | |
BI460301.1 | 2 | 135 | ||
BI550657.1 | 2 | 145 | ||
BI551732.1 | 2 | 118 | ||
BU661163.1 | ||||
CB991926.1 | 2 | 158 | ||
DA178808.1 | 2 | 113 | ||
DB103294.1 | ||||
DB494037.1 | 2 | 145 | ||
HY004717.1 | 2 | 170 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv435664 | Chr.16:15247194 - 18197960 on Build GRCh38 | Deletion | LOC102723692 NPIPA7 MIR6511A3 MYH11 MIR3670-2 XYLT1 NOMO3 PKD1P1 NDE1 MIR6511A2 MPV17L ABCC6 MIR6770-2 MIR484 MIR3179-2 ABCC1 MIR3180-2 NPIPA5 C16orf45 MIR6506 KIAA0430 PKD1P2 FOPNL |
dgv4926n54 | Chr.16:15883875 - 15924732 on Build GRCh38 | Gain | FOPNL |
nsv471085 | Chr.16:15847168 - 15931893 on Build GRCh38 | Gain | MYH11 FOPNL |
nsv833149 | Chr.16:15815546 - 15979954 on Build GRCh38 | Loss | ABCC1 MYH11 FOPNL |
nsv514795 | Chr.16:15883901 - 15884296 on Build GRCh38 | Gain | FOPNL |
esv3638042 | Chr.16:15877962 - 15918615 on Build GRCh38 | Gain | FOPNL |
esv2714015 | Chr.16:15292092 - 18247911 on Build GRCh38 | Deletion | LOC102723692 NPIPA7 MIR6511A3 MYH11 MIR3670-2 XYLT1 NOMO3 PKD1P1 NDE1 MIR6511A2 MPV17L ABCC6 MIR6770-2 MIR484 MIR3179-2 ABCC1 MIR3180-2 NPIPA5 C16orf45 MIR6506 KIAA0430 PKD1P2 FOPNL |
nsv1048188 | Chr.16:15874385 - 15911711 on Build GRCh38 | Gain | FOPNL |
esv23840 | Chr.16:15881982 - 15930025 on Build GRCh38 | Gain+Loss | FOPNL |
nsv1047898 | Chr.16:15388065 - 18284986 on Build GRCh38 | Loss | LOC102723692 NPIPA7 MIR6511A3 MYH11 MIR3670-2 XYLT1 NOMO3 PKD1P1 NDE1 MIR6511A2 MPV17L ABCC6 MIR6770-2 MIR484 MIR3179-2 ABCC1 MIR3180-2 C16orf45 MIR6506 KIAA0430 PKD1P2 FOPNL |
nsv457428 | Chr.16:15386022 - 18071186 on Build GRCh38 | Gain | LOC102723692 NPIPA7 MIR6511A3 MYH11 MIR3670-2 XYLT1 NOMO3 PKD1P1 NDE1 MIR6511A2 MPV17L ABCC6 MIR6770-2 MIR484 MIR3179-2 ABCC1 MIR3180-2 C16orf45 MIR6506 KIAA0430 PKD1P2 FOPNL |
nsv1125134 | Chr.16:15180201 - 18091718 on Build GRCh38 | Deletion | LOC102723692 NPIPA7 MIR6511A3 MYH11 MIR3670-2 XYLT1 NOMO3 PKD1P1 NDE1 MIR6511A2 MPV17L ABCC6 MIR6770-2 MIR484 MIR3179-2 ABCC1 MIR3180-2 NPIPA5 C16orf45 MIR6506 KIAA0430 PKD1P2 FOPNL |
esv2421345 | Chr.16:15882156 - 15931806 on Build GRCh38 | Duplication | FOPNL |
esv3638041 | Chr.16:15702756 - 16002857 on Build GRCh38 | Gain | ABCC1 MYH11 NDE1 FOPNL |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |