Assay Details
Target Gene Details
Entrez Gene ID: | 11073 |
Gene Name: | topoisomerase (DNA) II binding protein 1 |
Gene Aliases: |
TOP2BP1 |
Location: |
Chr.3:133600605-133662380 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOPBP1 | NM_007027.3 | 20 | 3403 | NP_008958.2 |
XM_005247076.3 | 20 | 3875 | XP_005247133.1 | |
XM_011512357.2 | 20 | 3474 | XP_011510659.1 | |
XM_017005636.1 | 20 | 3473 | XP_016861125.1 | |
XM_017005637.1 | 17 | 2927 | XP_016861126.1 | |
AB019397.1 | 20 | 3350 | BAA34202.1 | |
AK302584.1 | 20 | 3369 | ||
BC126209.1 | 19 | 3293 | ||
BC151237.1 | 20 | 3356 | ||
D87448.1 | 20 | 3356 | ||
Y18557.1 | 5 | 496 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |