Assay Details
Target Gene Details
Entrez Gene ID: | 5152 |
Gene Name: | phosphodiesterase 9A |
Gene Aliases: |
HSPDE9A2 |
Location: |
Chr.21:42653752-42775509 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3534 | Chr.21:42650074 - 42681450 on Build GRCh38 | Insertion | PDE9A LOC101928255 |
nsv1112803 | Chr.21:42653390 - 42654190 on Build GRCh38 | Deletion | PDE9A LOC101928255 |
nsv522001 | Chr.21:42421796 - 42827928 on Build GRCh38 | Gain | LOC101928311 PDE9A LOC101928255 LOC101928212 SLC37A1 RSPH1 LOC101928284 UBASH3A LOC101928233 LOC101930094 |
nsv587654 | Chr.21:42631745 - 42665548 on Build GRCh38 | Loss | PDE9A LOC101928255 |
esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion | WDR4 CBS LOC101928255 PKNOX1 NDUFV3 LOC400870 HSF2BP CRYAA U2AF1 LINC00319 LOC101928311 SIK1 LINC00313 PDE9A ERVH48-1 FRGCA MIR5692B LOC101928284 LINC00322 TCONS_00029157 |
nsv953634 | Chr.21:42648991 - 42669590 on Build GRCh38 | Deletion | PDE9A LOC101928255 |
nsv1073134 | Chr.21:42653389 - 42654190 on Build GRCh38 | Deletion | PDE9A LOC101928255 |
esv3647086 | Chr.21:42641558 - 42685014 on Build GRCh38 | Gain | PDE9A LOC101928255 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |