Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL1L | NM_001307976.1 | 14 | 3105 | NP_001294905.1 |
NM_152519.3 | 15 | 3231 | NP_689732.2 | |
XM_005246328.4 | 15 | 3513 | XP_005246385.1 | |
XM_005246329.4 | 15 | 3219 | XP_005246386.1 | |
XM_005246330.4 | 15 | 3100 | XP_005246387.1 | |
XM_005246332.4 | 14 | 3386 | XP_005246389.1 | |
XM_011510706.2 | 15 | 3138 | XP_011509008.1 | |
XM_011510707.2 | 15 | 3321 | XP_011509009.1 | |
XM_011510710.2 | 14 | 1970 | XP_011509012.1 | |
XM_017003430.1 | 15 | 3257 | XP_016858919.1 | |
XM_017003431.1 | 11 | 5339 | XP_016858920.1 | |
AK074441.1 | 14 | 2000 | BAB85085.1 | |
AL133053.1 | 9 | 1223 | CAB61379.1 | |
BC126155.1 | 14 | 3011 | ||
BC143914.1 | 14 | 3011 |
Target Gene Details
Entrez Gene ID: | 6120 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
RPE2-1 |
Location: |
Chr.2:210002565-210029156 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPE | XM_006712677.3 | 7 | 3873 | XP_006712740.1 |
AK055724.1 | 1 | 527 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |