Assay Details
Target Gene Details
Entrez Gene ID: | 50804 |
Gene Name: | myelin expression factor 2 |
Gene Aliases: |
HsT18564, MEF-2, MST156, MSTP156, myEF-2 |
Location: |
Chr.15:48134632-48178517 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYEF2 | NM_001301210.1 | 16 | 2679 | NP_001288139.1 |
NM_016132.4 | 17 | 2751 | NP_057216.2 | |
XM_005254422.4 | 16 | 2856 | XP_005254479.2 | |
XM_005254424.4 | 15 | 2598 | XP_005254481.2 | |
XM_005254425.4 | 16 | 2855 | XP_005254482.2 | |
XM_006720553.3 | 16 | 2699 | XP_006720616.1 | |
XM_011521657.2 | 13 | 2249 | XP_011519959.1 | |
XM_017022285.1 | 15 | 2804 | XP_016877774.1 | |
XM_017022286.1 | 15 | 2783 | XP_016877775.1 | |
XM_017022287.1 | 14 | 2732 | XP_016877776.1 | |
XM_017022288.1 | 15 | 2648 | XP_016877777.1 | |
XM_017022289.1 | 15 | 2627 | XP_016877778.1 | |
XM_017022290.1 | 14 | 2576 | XP_016877779.1 | |
XM_017022291.1 | 14 | 2575 | XP_016877780.1 | |
XM_017022292.1 | 13 | 2524 | XP_016877781.1 | |
AB037762.1 | 17 | 2689 | ||
AF190159.1 | 16 | 2442 | AAQ13703.1 | |
AK002075.1 | 5 | 1727 | BAA92070.1 | |
BC014533.1 | 6 | 1466 | AAH14533.3 | |
BC152420.1 | 17 | 2689 | ||
BC152451.1 | 17 | 2689 | ||
BX647197.1 | 3 | 1864 |
Target Gene Details
Entrez Gene ID: | 283652 |
Gene Name: | solute carrier family 24 member 5 |
Gene Aliases: |
JSX, NCKX5, OCA6, SHEP4 |
Location: |
Chr.15:48120972-48142392 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC24A5 | NM_205850.2 | 9 | 1309 | NP_995322.1 |
AF348468.1 | 9 | 1309 | AAQ15116.1 | |
BC073944.1 | 4 | 390 | AAH73944.1 | |
BC113628.1 | 9 | 1264 | ||
BC113630.1 | 9 | 1264 | ||
BC143950.1 | 9 | 1264 | ||
DQ665306.1 | 9 | 1257 | ||
DQ665307.1 | 8 | 1077 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832999 | Chr.15:47960097 - 48160208 on Build GRCh38 | Gain | MYEF2 SLC24A5 |
nsv569338 | Chr.15:48104611 - 48160602 on Build GRCh38 | Loss | MYEF2 SLC24A5 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |