Assay Details
Target Gene Details
Entrez Gene ID: | 55911 |
Gene Name: | apolipoprotein B receptor |
Gene Aliases: |
APOB100R, APOB48R |
Location: |
Chr.16:28494649-28498970 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOBR | NM_018690.3 | 2 | 1498 | NP_061160.3 |
AF141332.1 | 3 | 1448 | AAF76255.1 | |
AF141334.1 | 3 | 1448 | AAF78765.1 | |
BC119786.1 | 3 | 1448 | ||
BC119788.1 | 3 | 1243 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509608 | Chr.16:28449797 - 28632895 on Build GRCh38 | Insertion | SULT1A2 CLN3 APOBR NUPR1 SGF29 SULT1A1 IL27 |
esv2758640 | Chr.16:28163817 - 29139232 on Build GRCh38 | Gain+Loss | NPIPB8 SULT1A2 ATXN2L SBK1 XPO6 NPIP SH2B1 SGF29 SULT1A1 MIR6862-1 CLN3 ATP2A1 NPIPB9 IL27 TUFM LOC101928215 MIR4517 NPIPB6 NFATC2IP MIR6862-2 RABEP2 APOBR NUPR1 CD19 EIF3CL MIR4721 RRN3P2 ATP2A1-AS1 LAT EIF3C SPNS1 |
nsv1059555 | Chr.16:28373144 - 29195838 on Build GRCh38 | Gain | NPIPB8 SULT1A2 ATXN2L NPIP SH2B1 SGF29 LOC107984832 SULT1A1 MIR6862-1 CLN3 ATP2A1 NPIPB9 IL27 TUFM LOC101928215 MIR4517 NFATC2IP MIR6862-2 RABEP2 APOBR NUPR1 CD19 EIF3CL MIR4721 RRN3P2 ATP2A1-AS1 LAT EIF3C SPNS1 |
nsv833180 | Chr.16:28346157 - 28574165 on Build GRCh38 | Loss | MIR6862-1 NPIPB6 CLN3 APOBR NUPR1 SGF29 EIF3CL IL27 |
nsv9415 | Chr.16:28468881 - 28498959 on Build GRCh38 | Gain | CLN3 APOBR |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |