Assay Details
Target Gene Details
Entrez Gene ID: | 171017 |
Gene Name: | zinc finger protein 384 |
Gene Aliases: |
CAGH1, CAGH1A, CIZ, ERDA2, NMP4, NP, TNRC1 |
Location: |
Chr.12:6666477-6689572 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss | ZNF384 ING4 PIANP COPS7A SCARNA11 MLF2 ACRBP LPAR5 CHD4 |
nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss | ZNF384 USP5 P3H3 COPS7A GPR162 LAG3 ING4 PTMS PIANP CD4 CDCA3 MLF2 GNB3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |