Assay Details
Target Gene Details
Entrez Gene ID: | 2954 |
Gene Name: | glutathione S-transferase zeta 1 |
Gene Aliases: |
GSTZ1-1, MAAI, MAI |
Location: |
Chr.14:77320887-77331597 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSTZ1 | NM_001312660.1 | 6 | 767 | NP_001299589.1 |
NM_145870.2 | 5 | 581 | NP_665877.1 | |
NM_145871.2 | NP_665878.2 | |||
XM_011536670.2 | 5 | 1182 | XP_011534972.1 | |
XM_011536671.2 | XP_011534973.1 | |||
XM_017021243.1 | 4 | 852 | XP_016876732.1 | |
AB209360.1 | 2 | 4707 | BAD92597.1 | |
AJ001838.1 | 5 | 402 | CAA05045.1 | |
AK309054.1 | 4 | 788 | ||
AK315154.1 | 6 | 658 | ||
AK315609.1 | 5 | 425 | ||
BC001453.1 | 5 | 457 | AAH01453.1 | |
BG330365.1 | ||||
BG772450.1 | 5 | 588 | ||
BI559916.1 | 6 | 676 | ||
BI829992.1 | 6 | 647 | ||
CR456987.1 | 4 | 299 | CAG33268.1 | |
U86529.1 | 5 | 402 | AAB96392.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |