Assay Details
Target Gene Details
Entrez Gene ID: | 80830 |
Gene Name: | apolipoprotein L6 |
Gene Aliases: |
APOL-VI, APOLVI |
Location: |
Chr.22:35648317-35668409 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOL6 | NM_030641.3 | 3 | 1395 | NP_085144.1 |
XM_011530392.2 | 4 | 1572 | XP_011528694.1 | |
AK074645.1 | 1 | 883 | BAC11109.1 | |
AK095881.1 | 3 | 1357 | ||
AL834420.1 | 3 | 1359 | CAH56356.1 | |
AX775805.1 | 3 | 1367 | CAE11602.1 | |
AY014879.1 | 3 | 1395 | AAK07724.1 | |
AY358210.1 | 3 | 1292 | AAQ88577.1 | |
BC038950.2 | 3 | 1325 | AAH38950.2 | |
BC047864.1 | 3 | 1377 | AAH47864.1 | |
CR456381.1 | CAG30267.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |