Assay Details
Target Gene Details
Entrez Gene ID: | 2208 |
Gene Name: | Fc fragment of IgE receptor II |
Gene Aliases: |
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF |
Location: |
Chr.19:7688757-7702755 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCER2 | NM_001207019.2 | NP_001193948.2 | ||
NM_001220500.1 | NP_001207429.1 | |||
NM_002002.4 | NP_001993.2 | |||
XM_005272462.4 | XP_005272519.1 | |||
AK223207.1 | BAD96927.1 | |||
AK310071.1 | ||||
AK310707.1 | 8 | 636 | ||
BC014108.2 | AAH14108.1 | |||
BC062591.1 | AAH62591.1 | |||
M14766.1 | AAA52435.1 | |||
M15059.1 | AAA52434.1 | |||
X04772.1 | CAA28465.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953959 | Chr.19:7669315 - 7700414 on Build GRCh38 | Deletion | FCER2 MCEMP1 TRAPPC5 RETN |
nsv833731 | Chr.19:7589114 - 7739331 on Build GRCh38 | Loss | PET100 XAB2 CLEC4G CAMSAP3 FCER2 PCP2 MCEMP1 MIR6792 TRAPPC5 RETN STXBP2 |
nsv1060887 | Chr.19:7089546 - 7861087 on Build GRCh38 | Gain | PET100 ZNF358 INSR XAB2 PEX11G CLEC4G PNPLA6 FCER2 C19orf45 PCP2 ARHGEF18 MCEMP1 EVI5L MCOLN1 CLEC4M LOC107985283 CLEC4GP1 LOC100128573 CAMSAP3 MIR6792 TRAPPC5 RETN CD209 STXBP2 |
nsv833732 | Chr.19:7672656 - 7871329 on Build GRCh38 | Gain | PRR36 CLEC4G FCER2 MCEMP1 EVI5L TRAPPC5 CD209 CLEC4M CLEC4GP1 |
esv2751809 | Chr.19:6896369 - 7816144 on Build GRCh38 | Gain | MBD3L2 ADGRE1 XAB2 CLEC4G C19orf45 PCP2 ARHGEF18 MCEMP1 MCOLN1 CLEC4GP1 CAMSAP3 MBD3L5 MBD3L3 RETN PET100 ZNF358 INSR ADGRE4P PEX11G PNPLA6 FCER2 ZNF557 CLEC4M LOC107985283 MBD3L4 LOC100128573 FLJ25758 MIR6792 TRAPPC5 CD209 STXBP2 LOC729458 |
esv26340 | Chr.19:7690078 - 7698176 on Build GRCh38 | Gain | FCER2 |
nsv1061452 | Chr.19:7657750 - 7785131 on Build GRCh38 | Gain | CLEC4G FCER2 MCEMP1 TRAPPC5 RETN CD209 CLEC4M |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |