Assay Details
Target Gene Details
Entrez Gene ID: | 57212 |
Gene Name: | TP73 antisense RNA 1 |
Gene Aliases: |
KIAA0495, PDAM |
Location: |
Chr.1:3735984-3747373 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP73-AS1 | NR_033708.1 | 1 | 825 | |
NR_033709.1 | 1 | 825 | ||
NR_033710.1 | 1 | 825 | ||
NR_033711.1 | 1 | 825 | ||
NR_033712.1 | 1 | 223 | ||
AB007964.1 | 1 | 774 | ||
AL133648.1 | 1 | 78 | CAB63767.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv460061 | Chr.1:3680453 - 3755227 on Build GRCh38 | Loss |
![]() ![]() ![]() |
nsv470687 | Chr.1:3356499 - 3763210 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1001715 | Chr.1:3627619 - 3769754 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830425 | Chr.1:3740889 - 3841825 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv951959 | Chr.1:3051037 - 3812036 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv830314 | Chr.1:3660776 - 3804275 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |
dgv13n100 | Chr.1:3730448 - 3791316 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)