Assay Details
Target Gene Details
Entrez Gene ID: | 55152 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:49015399-49021386 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DALRD3 | NM_001009996.2 | NP_001009996.1 | ||
NM_001276405.1 | NP_001263334.1 | |||
NM_018114.5 | NP_060584.3 | |||
XM_005265269.3 | XP_005265326.1 | |||
XM_006713219.2 | XP_006713282.1 | |||
XM_011533891.1 | 10 | 1594 | XP_011532193.1 | |
XM_011533892.1 | XP_011532194.1 | |||
XM_011533893.1 | 8 | 1603 | XP_011532195.1 | |
XM_017006723.1 | XP_016862212.1 | |||
AA553582.1 | 1 | 72 | ||
AK001358.1 | 8 | 1669 | BAA91647.1 | |
AK093054.1 | 2 | 2631 | ||
AK093204.1 | BAC04095.1 | |||
AK093294.1 | BAC04123.1 | |||
AK308273.1 | ||||
BC032440.1 | AAH32440.2 | |||
BC047683.1 | AAH47683.1 | |||
BC054493.1 | AAH54493.1 | |||
BX648719.1 | 6 | 1269 |
Target Gene Details
Entrez Gene ID: | 11180 |
Gene Name: | WD repeat domain 6 |
Gene Aliases: |
- |
Location: |
Chr.3:49007062-49015953 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WDR6 | NM_001320546.1 | 6 | 4509 | NP_001307475.1 |
NM_001320547.1 | 6 | 3880 | NP_001307476.1 | |
NM_018031.4 | 6 | 4295 | NP_060501.3 | |
AF099100.1 | 6 | 3964 | AAF80244.1 | |
AI636120.1 | 1 | 121 | ||
AK001080.1 | BAA91497.1 | |||
AK022719.1 | 5 | 2200 | BAB14201.1 | |
AK092079.1 | 6 | 1620 | ||
AK225653.1 | ||||
AL133589.1 | 5 | 2075 | CAB63730.1 | |
AL833910.1 | 5 | 3141 | CAD38766.1 | |
BC002826.2 | 8 | 1976 | AAH02826.1 | |
BC078176.1 | 5 | 2014 | AAH78176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv954488 | Chr.3:49002068 - 49019667 on Build GRCh38 | Deletion | P4HTM WDR6 DALRD3 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |