Assay Details
Target Gene Details
Entrez Gene ID: | 527 |
Gene Name: | ATPase H+ transporting V0 subunit c |
Gene Aliases: |
ATP6C, ATP6L, ATPL, VATL, VPPC, Vma3 |
Location: |
Chr.16:2513726-2520223 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ATP6V0C | NM_001198569.1 | NP_001185498.1 | ||
NM_001694.3 | 1 | 95 | NP_001685.1 | |
AW161158.1 | ||||
BC004537.2 | AAH04537.1 | |||
BC007389.2 | AAH07389.1 | |||
BC007759.2 | AAH07759.1 | |||
BC009290.2 | AAH09290.1 | |||
BI548787.1 | ||||
M62762.1 | 1 | 95 | AAA60039.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication | LOC652276 MIR3178 NPRL3 NDUFB10 TSR3 MIR4516 C16orf59 SYNGR3 HS3ST6 FAM234A TMEM8A ZNF598 STUB1 TSC2 PIGQ WASIR2 TELO2 HBA1 MIR6511B1 UNKL NTN3 CASKIN1 SOX8 MLST8 HN1L SLC9A3R2 PKD1 LMF1 METRN BRICD5 RHBDL1 AXIN1 HBM LINC00254 WDR90 MIR6859-4 LOC100287175 RHBDF1 LOC105371046 LOC105371038 CHTF18 FBXL16 ABCA3 PTX4 MIR5587 MAPK8IP3 HBA2 NHLRC4 MIR6768 HAGHL IGFALS CEMP1 PRSS29P JMJD8 TPSB2 RNPS1 C1QTNF8 NPW CACNA1H MCRIP2 RPS2 E4F1 CCDC78 MIR940 NOXO1 RPUSD1 SNORA64 TPSG1 PRSS27 RAB40C SPSB3 NTHL1 TBL3 PDIA2 SNRNP25 MIR4717 MPG LOC100134368 PDPK1 FLJ42627 LOC101929280 PRR35 HBZ C16orf91 LOC107984876 ABCA17P PGP MIR3677 FAHD1 CRAMP1 SNHG9 POLR3K WFIKKN1 NME3 LOC729652 IFT140 LMF1-AS1 HBQ1 TMEM204 KCTD5 RNF151 GNPTG GNG13 SNORA78 LOC106660606 CCNF CAPN15 FAM173A RAB11FIP3 MSRB1 MIR3177 LUC7L UBE2I WDR24 HAGH DNASE1L2 ERVK13-1 NARFL BAIAP3 EME2 NME4 MIR3176 ECI1 SNHG19 MIR3180-5 RPL3L MEIOB METTL26 SSTR5 RPL23AP5 LINC00235 CCDC154 TRAF7 TPSD1 MRPL28 LOC105371045 SNORD60 LOC105371184 MIR1225 DDX11L10 SSTR5-AS1 ATP6V0C GFER RGS11 RHOT2 MRPS34 RAB26 MIR662 NUBP2 PRR25 ARHGDIG MIR6767 DECR2 AMDHD2 LOC107987233 TPSAB1 LOC105371049 TBC1D24 LOC105371047 SNORA10 MSLN CLCN7 |
esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss | LOC652276 MIR3178 PRSS27 CEMP1 AMDHD2 ATP6V0C SRRM2-AS1 ERVK13-1 TBC1D24 SRRM2 PDPK1 FLJ42627 TCEB2 PRSS33 PRSS41 KCTD5 PRSS21 |
nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss | LOC652276 MIR3178 ERVK13-1 CEMP1 TBC1D24 PDPK1 FLJ42627 AMDHD2 ATP6V0C |
nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain | LOC652276 MIR6768 MIR3178 TRAF7 ABCA17P MIR4516 CEMP1 C16orf59 SNORD60 RNPS1 PGP MIR1225 ATP6V0C MIR3677 TSC2 E4F1 MIR6511B1 NTN3 CASKIN1 LOC729652 MLST8 MIR940 RAB26 PKD1 BRICD5 LOC106660606 MIR6767 CCNF AMDHD2 LOC105371049 MIR4717 DNASE1L2 TBC1D24 PDPK1 ABCA3 ECI1 SNHG19 MIR3180-5 |
nsv833121 | Chr.16:2360975 - 2518228 on Build GRCh38 | Loss | MIR6768 NTN3 ABCA17P MIR6767 C16orf59 TBC1D24 CCNF LOC729652 ATP6V0C |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |