Assay Details
Target Gene Details
Entrez Gene ID: | 10146 |
Gene Name: | G3BP stress granule assembly factor 1 |
Gene Aliases: |
G3BP, HDH-VIII |
Location: |
Chr.5:151771915-151811352 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
G3BP1 | NM_005754.2 | NP_005745.1 | ||
NM_198395.1 | NP_938405.1 | |||
XM_006714749.3 | XP_006714812.1 | |||
XM_006714750.3 | XP_006714813.1 | |||
XM_017008922.1 | XP_016864411.1 | |||
XM_017008923.1 | XP_016864412.1 | |||
AK130003.1 | BAC85272.1 | |||
AK222606.1 | BAD96326.1 | |||
AK300098.1 | ||||
AK303580.1 | ||||
AK309172.1 | ||||
AK312572.1 | ||||
BC006997.1 | AAH06997.1 | |||
BC108278.1 | AAI08279.1 | |||
BG703413.1 | ||||
BT019384.1 | AAV38191.1 | |||
BU517281.1 | ||||
BX647869.1 | CAI46065.1 | |||
BX648012.1 | CAI46067.1 | |||
CR536535.1 | CAG38772.1 | |||
U32519.1 | AAB07787.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |