Assay Details
Target Gene Details
Entrez Gene ID: | 4701 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A7 |
Gene Aliases: |
B14.5a, CI-B14.5a |
Location: |
Chr.19:8308603-8321413 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA7 | NM_005001.4 | 1 | 45 | NP_004992.2 |
NR_135539.1 | 1 | 45 | ||
AF050637.1 | AAD05427.1 | |||
AF054178.1 | AAC99399.1 | |||
BC003102.1 | AAH03102.1 | |||
BC107892.1 | AAI07893.1 | |||
CD172691.1 | 1 | 79 | ||
CR456915.1 | CAG33196.1 | |||
CR542266.1 | CAG47062.1 | |||
CR975510.1 | ||||
DA189271.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |