Assay Details
Target Gene Details
Entrez Gene ID: | 3383 |
Gene Name: | intercellular adhesion molecule 1 |
Gene Aliases: |
BB2, CD54, P3.58 |
Location: |
Chr.19:10270841-10286615 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ICAM1 | NM_000201.2 | 2 | 543 | NP_000192.2 |
AF340038.1 | 2 | 224 | AAQ14901.1 | |
AF340039.1 | 2 | 212 | AAQ14902.1 | |
AK130659.1 | 2 | 264 | ||
AK298054.1 | 2 | 150 | ||
AK298983.1 | ||||
AK301412.1 | ||||
AK312636.1 | 2 | 264 | ||
BC015969.2 | 2 | 295 | AAH15969.1 | |
BP346334.1 | ||||
BT006854.1 | 2 | 224 | AAP35500.1 | |
J03132.1 | 2 | 281 | AAA52709.1 | |
M24283.1 | 2 | 295 | AAA52708.1 | |
M55024.1 | AAA60392.1 | |||
X06990.1 | 2 | 236 | CAA30051.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv578612 | Chr.19:10271961 - 10284355 on Build GRCh38 | Gain | ICAM1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |