Assay Details
Target Gene Details
Entrez Gene ID: | 28952 |
Gene Name: | coiled-coil domain containing 22 |
Gene Aliases: |
CXorf37, JM1, RTSC2 |
Location: |
Chr.X:49235467-49250526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC22 | NM_014008.4 | NP_054727.1 | ||
XM_005272599.3 | XP_005272656.1 | |||
AJ005890.1 | CAA06747.1 | |||
AK291976.1 | ||||
AK296911.1 | ||||
AK298264.1 | ||||
AK316375.1 | ||||
BC000972.2 | AAH00972.1 | |||
BC011675.2 | AAH11675.1 | |||
BG912845.1 | ||||
DC340647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv471602 | Chr.X:49205142 - 49320261 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv469734 | Chr.X:49205141 - 49320261 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() |
nsv6895 | Chr.X:49227319 - 49262690 on Build GRCh38 | Insertion |
![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)