Assay Details
Target Gene Details
Entrez Gene ID: | 23389 |
Gene Name: | mediator complex subunit 13 like |
Gene Aliases: |
MRFACD, PROSIT240, THRAP2, TRAP240L |
Location: |
Chr.12:115958576-116277219 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MED13L | NM_015335.4 | 31 | 6802 | NP_056150.1 |
XM_011538080.2 | 31 | 6871 | XP_011536382.1 | |
XM_011538081.2 | 31 | 6868 | XP_011536383.1 | |
XM_011538082.2 | 31 | 7041 | XP_011536384.1 | |
XM_017019090.1 | 31 | 6832 | XP_016874579.1 | |
AB028948.2 | 25 | 5869 | BAA82977.2 | |
AF515599.1 | 31 | 6802 | AAQ08182.1 | |
AK023837.1 | 15 | 3519 | BAB14697.1 | |
AL133033.1 | 25 | 5842 | CAB61363.1 | |
AL137644.1 | 8 | 1345 | CAB70855.1 | |
AY338463.1 | 31 | 6954 | AAR08418.1 | |
BC130422.1 | 31 | 6798 | ||
CR749332.1 | 29 | 6326 | CAH18186.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |