Assay Details
Target Gene Details
Entrez Gene ID: | 377841 |
Gene Name: | ectonucleoside triphosphate diphosphohydrolase 8 |
Gene Aliases: |
E-NTPDase, GLSR2492, NTPDase-8, UNQ2492 |
Location: |
Chr.9:137434364-137441710 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ENTPD8 | NM_001033113.1 | NP_001028285.1 | ||
NM_198585.2 | NP_940987.2 | |||
XM_011518665.2 | XP_011516967.1 | |||
XM_011518666.2 | XP_011516968.1 | |||
XM_011518668.1 | XP_011516970.1 | |||
AY359088.1 | AAQ89446.1 | |||
AY430414.1 | AAR04374.1 | |||
AY903953.1 | AAW83515.1 | |||
AY903954.1 | AAW83516.1 | |||
BC141810.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv616021 | Chr.9:137434689 - 137452821 on Build GRCh38 | Loss | NSMF MIR7114 ENTPD8 |
nsv1049808 | Chr.9:137356366 - 137438026 on Build GRCh38 | Gain | EXD3 ENTPD8 NOXA1 |
nsv951208 | Chr.9:137379349 - 137467948 on Build GRCh38 | Deletion | EXD3 NSMF MIR7114 ENTPD8 PNPLA7 NOXA1 |
nsv6771 | Chr.9:137412670 - 137444666 on Build GRCh38 | Insertion | EXD3 ENTPD8 NOXA1 |
esv2759721 | Chr.9:137303332 - 137447299 on Build GRCh38 | Loss | EXD3 ENTPD8 NOXA1 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | PRR31 GRIN1 TMEM203 RABL6 SLC34A3 NDOR1 LOC101930307 EXD3 CCDC183 ANAPC2 TRAF2 NRARP MIR7114 PHPT1 RNF224 MAN1B1 NOXA1 FAM166A C8G C9orf172 MIR3621 ENTPD8 FBXW5 LRRC26 MIR4479 RNF208 NSMF TPRN MIR4292 UAP1L1 C9orf142 TMEM210 NELFB LCNL1 ABCA2 C9orf139 MAMDC4 EDF1 CLIC3 TUBB4B CCDC183-AS1 STPG3 C9orf173-AS1 NPDC1 PNPLA7 PTGDS SAPCD2 MAN1B1-AS1 SSNA1 FUT7 TOR4A ENTPD2 CYSRT1 DPP7 LCN12 |
esv3622034 | Chr.9:137433077 - 137439026 on Build GRCh38 | Gain | ENTPD8 NOXA1 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | NELFB GRIN1 TMEM203 SLC34A3 FAM166A STPG3 C9orf173-AS1 MIR3621 ENTPD8 NDOR1 LRRC26 EXD3 ANAPC2 SSNA1 NRARP TOR4A RNF208 TPRN CYSRT1 TUBB4B TMEM210 RNF224 NOXA1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |