Assay Details
Target Gene Details
Entrez Gene ID: | 256281 |
Gene Name: | nudix hydrolase 14 |
Gene Aliases: |
UGPP, UGPPase |
Location: |
Chr.14:105172937-105181323 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NUDT14 | NM_177533.4 | NP_803877.2 | ||
AB087802.1 | BAC65455.1 | |||
AK128196.1 | ||||
BC041584.1 | AAH41584.1 | |||
BE671270.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv566156 | Chr.14:105125040 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv566177 | Chr.14:105167384 - 105177639 on Build GRCh38 | Gain | JAG2 NUDT14 |
esv2760014 | Chr.14:105047009 - 105483089 on Build GRCh38 | Loss | MTA1 BTBD6 JAG2 LOC102723354 BRF1 CRIP2 MIR6765 PACS2 LOC100507437 GPR132 NUDT14 TEX22 |
nsv470679 | Chr.14:105124547 - 105605978 on Build GRCh38 | Loss | LOC105370697 MTA1 JAG2 CRIP2 CRIP1 IGHA2 LOC100507437 NUDT14 TMEM121 IGH C14orf80 BTBD6 BRF1 MIR6765 PACS2 TEX22 |
nsv9175 | Chr.14:105030172 - 105222604 on Build GRCh38 | Gain | JAG2 LOC102723354 BRF1 MIR6765 GPR132 NUDT14 |
nsv952279 | Chr.14:105136864 - 105237863 on Build GRCh38 | Deletion | JAG2 BRF1 MIR6765 NUDT14 |
nsv817646 | Chr.14:105149311 - 105188269 on Build GRCh38 | Loss | JAG2 MIR6765 NUDT14 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | LOC105370708 ZFYVE21 CDCA4 LINC00637 KIF26A JAG2 MIR203A LOC105370691 C14orf79 CEP170B SNORA28 TRMT61A ASPG NUDT14 AHNAK2 TMEM179 LOC107984648 LOC101929634 BTBD6 LINC00638 CKB GPR132 LOC105370687 C14orf2 MIR4710 INF2 TDRD9 LOC388022 AKT1 PPP1R13B MARK3 EIF5 LOC102723354 XRCC3 BAG5 PLD4 ADSSL1 SIVA1 C14orf180 KLC1 APOPT1 LOC105378183 RD3L MIR203B BRF1 MIR6765 PACS2 ZBTB42 |
nsv1052190 | Chr.14:104957424 - 105256316 on Build GRCh38 | Gain | AHNAK2 CDCA4 BTBD6 JAG2 LOC102723354 BRF1 C14orf79 MIR6765 GPR132 NUDT14 |
nsv517055 | Chr.14:105136282 - 106779068 on Build GRCh38 | Gain+Loss | MTA1 LOC107987211 ELK2AP MIR4537 IGHV3-41 JAG2 IGHV3-71 MIR8071-1 IGHG4 CRIP2 IGHV4-31 MIR8071-2 LOC100507437 NUDT14 IGHM TMEM121 IGH LINC00221 IGHG2 BTBD6 LINC00226 KIAA0125 IGHV3-48 IGHV3-69-1 TEX22 LOC105370697 IGHV4-55 IGHV3-30 CRIP1 IGHA2 IGHA1 IGHD C14orf80 IGHV3-23 MIR4507 ADAM6 BRF1 IGHV1-69 MIR6765 PACS2 IGHG1 MIR4539 IGHG3 MIR4538 |
nsv566150 | Chr.14:104969312 - 105223755 on Build GRCh38 | Loss | AHNAK2 CDCA4 JAG2 LOC102723354 BRF1 C14orf79 MIR6765 GPR132 NUDT14 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |