Assay Details
Target Gene Details
Entrez Gene ID: | 10011 |
Gene Name: | steroid receptor RNA activator 1 |
Gene Aliases: |
SRA, SRAP, STRAA1, pp7684 |
Location: |
Chr.5:140550067-140558093 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRA1 | NM_001035235.3 | NP_001030312.2 | ||
NM_001253764.1 | NP_001240693.1 | |||
NR_045586.1 | ||||
NR_045587.1 | ||||
AF092038.1 | ||||
AF293024.1 | AAG02114.1 | |||
AF293025.1 | AAG02115.1 | |||
AF293026.1 | AAG02116.1 | |||
AF318361.1 | AAL55868.1 | |||
AK024640.1 | ||||
AK054960.1 | ||||
BC040043.2 | AAH40043.2 | |||
BU553214.1 | ||||
CX165918.1 | ||||
DA311407.1 | ||||
DQ286291.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |