Assay Details
Target Gene Details
Entrez Gene ID: | 11047 |
Gene Name: | adhesion regulating molecule 1 |
Gene Aliases: |
ARM-1, ARM1, GP110 |
Location: |
Chr.20:62302056-62308862 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADRM1 | NM_001281437.1 | NP_001268366.1 | ||
NM_001281438.1 | NP_001268367.1 | |||
NM_007002.3 | NP_008933.2 | |||
NM_175573.2 | NP_783163.1 | |||
XM_005260257.1 | XP_005260314.1 | |||
XM_011528503.1 | XP_011526805.1 | |||
XM_017027602.1 | XP_016883091.1 | |||
XM_017027603.1 | XP_016883092.1 | |||
XM_017027604.1 | XP_016883093.1 | |||
XM_017027605.1 | XP_016883094.1 | |||
XM_017027606.1 | XP_016883095.1 | |||
XM_017027607.1 | XP_016883096.1 | |||
XM_017027608.1 | XP_016883097.1 | |||
XM_017027609.1 | XP_016883098.1 | |||
XM_017027610.1 | XP_016883099.1 | |||
BC010733.1 | AAH10733.1 | |||
BC017245.2 | AAH17245.1 | |||
CR749213.1 | CAH18070.2 | |||
D64154.1 | BAA11023.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2422398 | Chr.20:62207833 - 62674040 on Build GRCh38 | Duplication | SLCO4A1-AS1 MIR1-1 C20orf166-AS1 MIR133A2 HRH3 GATA5 MIR1-1HG RBBP8NL RPS21 MIR4758 LOC105369209 LAMA5 SLCO4A1 CABLES2 OSBPL2 LAMA5-AS1 ADRM1 |
nsv953301 | Chr.20:62305045 - 62338544 on Build GRCh38 | Deletion | LAMA5 MIR4758 ADRM1 |
nsv518296 | Chr.20:62203245 - 62415019 on Build GRCh38 | Loss | LAMA5 MTG2 CABLES2 HRH3 OSBPL2 RBBP8NL RPS21 LAMA5-AS1 MIR4758 ADRM1 LOC105369209 |
nsv3441 | Chr.20:62297903 - 62320059 on Build GRCh38 | Insertion | LAMA5 ADRM1 |
nsv586485 | Chr.20:62306724 - 62312525 on Build GRCh38 | Loss | LAMA5 ADRM1 |
nsv834024 | Chr.20:62259914 - 62455522 on Build GRCh38 | Gain | LAMA5 CABLES2 OSBPL2 RBBP8NL RPS21 LAMA5-AS1 MIR4758 ADRM1 |
nsv470556 | Chr.20:62212004 - 62577283 on Build GRCh38 | Loss | MIR1-1 C20orf166-AS1 MIR133A2 HRH3 GATA5 MIR1-1HG RBBP8NL RPS21 MIR4758 LOC105369209 LAMA5 CABLES2 OSBPL2 LAMA5-AS1 ADRM1 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |