Assay Details
Target Gene Details
Entrez Gene ID: | 10417 |
Gene Name: | spondin 2 |
Gene Aliases: |
DIL-1, DIL1, M-SPONDIN, MINDIN |
Location: |
Chr.4:1166932-1208962 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPON2 | NM_001128325.2 | NP_001121797.1 | ||
NM_001199021.1 | NP_001185950.1 | |||
NM_012445.3 | 1 | 172 | NP_036577.1 | |
AB027466.1 | 1 | 118 | BAA85892.1 | |
AK074618.1 | BAC11092.1 | |||
AK074770.1 | 1 | 110 | BAC11196.1 | |
AY358948.1 | 1 | 117 | AAQ89307.1 | |
BC036341.1 | 1 | 111 | AAH36341.1 | |
BY797606.2 | 1 | 156 | ||
DA087369.1 | ||||
DA092888.1 | ||||
DA201089.1 | ||||
DA375259.1 | 1 | 176 | ||
DB249869.1 | ||||
DB254983.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss | UVSSA TMED11P IDUA CTBP1 CTBP1-AS ATP5I CRIPAK LOC100130872 LOC105374344 LOC101928548 CPLX1 PCGF3 TMEM175 MFSD7 LOC101928521 RNF212 SPON2 LOC107986211 MAEA PDE6B DGKQ LOC100129917 CTBP1-AS2 FGFRL1 GAK MYL5 NKX1-1 SLC26A1 |
nsv593278 | Chr.4:1165892 - 1421910 on Build GRCh38 | Loss | LOC101928548 UVSSA MAEA CTBP1 CTBP1-AS SPON2 CTBP1-AS2 CRIPAK LOC100130872 NKX1-1 |
nsv950331 | Chr.4:1164113 - 1239212 on Build GRCh38 | Deletion | LOC101928548 CTBP1 CTBP1-AS SPON2 LOC100130872 |
nsv519106 | Chr.4:1165203 - 1181073 on Build GRCh38 | Loss | SPON2 |
nsv1012892 | Chr.4:1093083 - 1182842 on Build GRCh38 | Loss | LOC105374344 TMED11P RNF212 SPON2 |
nsv829835 | Chr.4:1103798 - 1310451 on Build GRCh38 | Gain | LOC105374344 LOC101928548 MAEA TMED11P CTBP1 RNF212 CTBP1-AS SPON2 CTBP1-AS2 LOC100130872 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |