Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | 2 | 3234 | NP_057356.1 |
XM_005261315.2 | 2 | 4445 | XP_005261372.1 | |
XM_006724105.3 | 2 | 3751 | XP_006724168.1 | |
XM_017028539.1 | 2 | 5527 | XP_016884028.1 | |
AB051450.1 | 2 | 2993 | ||
AL133080.1 | 1 | 136 | ||
BC038957.1 | 2 | 3007 | AAH38957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | PHF5A TOB2 RANGAP1 LOC100996598 ZC3H7B TEF |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | PHF5A TOB2 RANGAP1 PMM1 ACO2 LOC100996598 MIR6889 CSDC2 LOC105373044 ZC3H7B TEF POLR3H |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | PHF5A TOB2 ACO2 LOC100996598 LOC105373044 TEF POLR3H |
nsv962798 | Chr.22:41433542 - 41435164 on Build GRCh38 | Duplication | TOB2 |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | TOB2 RANGAP1 CHADL LOC100996598 MIR6889 ZC3H7B TEF |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | PHF5A C22orf46 TOB2 MEI1 EP300-AS1 LOC100996598 XRCC6 MIR6889 CSDC2 LOC105373044 RANGAP1 PMM1 EP300 ACO2 CHADL SNORD140 SNU13 DESI1 MIR1281 L3MBTL2 ZC3H7B TEF POLR3H |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |