Assay Details
Target Gene Details
Entrez Gene ID: | 55830 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
AD-017, MSTP139 |
Location: |
Chr.3:52694484-52706083 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLT8D1 | NM_001010983.2 | NP_001010983.1 | ||
NM_001278280.1 | NP_001265209.1 | |||
NM_001278281.1 | NP_001265210.1 | |||
NM_018446.3 | NP_060916.1 | |||
NM_152932.2 | NP_690909.1 | |||
XM_017006857.1 | XP_016862346.1 | |||
XM_017006858.1 | XP_016862347.1 | |||
XM_017006859.1 | XP_016862348.1 | |||
AF157318.1 | AAF67484.1 | |||
AF175227.1 | AAQ13652.1 | |||
AF176777.1 | AAQ13655.1 | |||
AJ271710.1 | CAC19666.1 | |||
AK027517.1 | BAB55170.1 | |||
AK075031.1 | 6 | 798 | BAC11363.1 | |
AY358579.1 | AAQ88942.1 | |||
BC119670.1 | ||||
BC119671.1 | ||||
CR749821.1 | CAH18681.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |