Assay Details
Target Gene Details
Entrez Gene ID: | 151050 |
Gene Name: | KAT8 regulatory NSL complex subunit 1 like |
Gene Aliases: |
C2orf67, MSL1v2 |
Location: |
Chr.2:210021421-210171370 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KANSL1L | NM_001307976.1 | 14 | 4375 | NP_001294905.1 |
NM_152519.3 | 15 | 4501 | NP_689732.2 | |
XM_005246328.4 | 15 | 4783 | XP_005246385.1 | |
XM_005246329.4 | 15 | 4489 | XP_005246386.1 | |
XM_005246330.4 | 15 | 4370 | XP_005246387.1 | |
XM_005246332.4 | 14 | 4656 | XP_005246389.1 | |
XM_011510706.2 | 15 | 4408 | XP_011509008.1 | |
XM_011510707.2 | 15 | 4591 | XP_011509009.1 | |
XM_011510710.2 | 14 | 3240 | XP_011509012.1 | |
XM_017003430.1 | 15 | 4527 | XP_016858919.1 | |
XM_017003431.1 | 11 | 6609 | XP_016858920.1 | |
AI289109.1 | 1 | 244 | ||
AI679696.1 | 1 | 123 | ||
AK074441.1 | 14 | 3270 | BAB85085.1 | |
BU568066.1 | 1 | 437 |
Target Gene Details
Entrez Gene ID: | 6120 |
Gene Name: | ribulose-5-phosphate-3-epimerase |
Gene Aliases: |
RPE2-1 |
Location: |
Chr.2:210002565-210029156 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
RPE | NM_001278282.1 | 7 | 2661 | NP_001265211.1 |
NM_001278283.1 | 8 | 2741 | NP_001265212.1 | |
NM_001278285.1 | 5 | 2569 | NP_001265214.1 | |
NM_001278286.1 | 6 | 2585 | NP_001265215.1 | |
NM_001278288.1 | 5 | 2576 | NP_001265217.1 | |
NM_001278289.1 | 7 | 2725 | NP_001265218.1 | |
NM_001318926.1 | 7 | 2710 | NP_001305855.1 | |
NM_001318927.1 | 7 | 2754 | NP_001305856.1 | |
NM_001318928.1 | 7 | 2605 | NP_001305857.1 | |
NM_001318929.1 | 6 | 2652 | NP_001305858.1 | |
NM_001318930.1 | 6 | 2645 | NP_001305859.1 | |
NM_001318931.1 | 6 | 2607 | NP_001305860.1 | |
NM_006916.2 | 6 | 2630 | NP_008847.1 | |
NM_199229.2 | 6 | 2656 | NP_954699.1 | |
XM_006712677.3 | 7 | 2603 | XP_006712740.1 | |
AB075523.1 | 1 | 1281 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |