Assay Details
Target Gene Details
Entrez Gene ID: | 5510 |
Gene Name: | protein phosphatase 1 regulatory subunit 7 |
Gene Aliases: |
SDS22 |
Location: |
Chr.2:241149573-241183650 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPP1R7 | NM_001282409.1 | NP_001269338.1 | ||
NM_001282410.1 | NP_001269339.1 | |||
NM_001282411.1 | NP_001269340.1 | |||
NM_001282412.1 | NP_001269341.1 | |||
NM_001282413.1 | NP_001269342.1 | |||
NM_001282414.1 | NP_001269343.1 | |||
NM_002712.2 | NP_002703.1 | |||
XM_011511388.1 | XP_011509690.1 | |||
AK294043.1 | ||||
AK308015.1 | ||||
BC000910.2 | AAH00910.1 | |||
BC012397.1 | AAH12397.1 | |||
BC013001.2 | AAH13001.1 | |||
BG684153.1 | ||||
BI256145.1 | ||||
BQ212238.1 | ||||
BT007296.1 | AAP35960.1 | |||
BT020134.1 | AAV38936.1 | |||
DA787059.1 | ||||
Z50749.1 | CAA90626.1 |
More Information
Set Membership: |
Intragenic Non-exonic non-DGV Variation |