Assay Details
Target Gene Details
Entrez Gene ID: | 347 |
Gene Name: | apolipoprotein D |
Gene Aliases: |
- |
Location: |
Chr.3:195568702-195584205 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
APOD | NM_001647.3 | 1 | 167 | NP_001638.1 |
AK294523.1 | ||||
BU195270.1 | ||||
DA892855.1 | 1 | 167 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5016n100 | Chr.3:195552380 - 195720039 on Build GRCh38 | Gain+Loss | LINC00969 MIR570 APOD SDHAP2 LOC105374297 |
nsv1001387 | Chr.3:195465410 - 195934667 on Build GRCh38 | Gain | LINC00969 MUC4 LOC101929697 LOC105374299 MUC20 PPP1R2 MIR570 APOD MIR6829 TNK2 SDHAP2 LOC105374297 |
dgv5015n100 | Chr.3:195532487 - 195751823 on Build GRCh38 | Loss | LINC00969 MUC4 MUC20 PPP1R2 MIR570 APOD SDHAP2 LOC105374297 |
nsv1006229 | Chr.3:195350692 - 195747880 on Build GRCh38 | Gain | LINC00969 MUC4 MUC20 PPP1R2 MIR570 ACAP2 APOD SDHAP2 LOC105374297 |
nsv428430 | Chr.3:195478549 - 196102555 on Build GRCh38 | Gain+Loss | LOC101929697 PPP1R2 MIR570 SDHAP1 MIR6829 TNK2 SDHAP2 TFRC LINC00969 MUC4 LOC105374301 LOC105374299 MUC20 APOD LOC105374297 |
esv2763776 | Chr.3:195514028 - 195775248 on Build GRCh38 | Gain+Loss | LINC00969 MUC4 MUC20 PPP1R2 MIR570 APOD SDHAP2 LOC105374297 |
dgv5014n100 | Chr.3:195514016 - 195747880 on Build GRCh38 | Gain | LINC00969 MUC4 MUC20 PPP1R2 MIR570 APOD SDHAP2 LOC105374297 |
esv2759213 | Chr.3:195382058 - 196344722 on Build GRCh38 | Gain+Loss | TCTEX1D2 LOC101929697 PPP1R2 MIR570 ACAP2 SDHAP1 MIR6829 TM4SF19-AS1 TNK2 SDHAP2 SLC51A TFRC PCYT1A LINC00885 LINC00969 MUC4 LOC105374301 LOC105374299 MUC20 ZDHHC19 APOD TM4SF19-TCTEX1D2 TM4SF19 LOC105374297 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |