Assay Details
Target Gene Details
Entrez Gene ID: | 6285 |
Gene Name: | S100 calcium binding protein B |
Gene Aliases: |
NEF, S100, S100-B, S100beta |
Location: |
Chr.21:46598618-46605242 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
S100B | NM_006272.2 | 3 | 904 | NP_006263.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv528495 | Chr.21:46595956 - 46632926 on Build GRCh38 | Loss | S100B |
dgv4442n100 | Chr.21:46551020 - 46677033 on Build GRCh38 | Gain | PRMT2 DIP2A S100B |
nsv587999 | Chr.21:46522003 - 46683939 on Build GRCh38 | Gain | PRMT2 DIP2A S100B |
nsv588002 | Chr.21:46575530 - 46653004 on Build GRCh38 | Gain | PRMT2 S100B |
esv34393 | Chr.21:46595759 - 46657900 on Build GRCh38 | Gain | PRMT2 S100B |
esv3647171 | Chr.21:46586178 - 46674643 on Build GRCh38 | Gain | PRMT2 S100B |
dgv4443n100 | Chr.21:46561965 - 46646565 on Build GRCh38 | Gain | PRMT2 DIP2A S100B |
esv2758822 | Chr.21:46566757 - 46657900 on Build GRCh38 | Gain | PRMT2 DIP2A S100B |
dgv4441n100 | Chr.21:46474792 - 46677033 on Build GRCh38 | Gain | PRMT2 DIP2A S100B |
nsv528947 | Chr.21:46566170 - 46630476 on Build GRCh38 | Gain | DIP2A S100B |
esv3647172 | Chr.21:46592879 - 46650385 on Build GRCh38 | Loss | PRMT2 S100B |
esv2760700 | Chr.21:46554142 - 46634864 on Build GRCh38 | Gain | DIP2A S100B |
nsv470910 | Chr.21:45933861 - 46665076 on Build GRCh38 | Gain | DIP2A-IT1 DIP2A COL6A2 MCM3AP S100B C21orf58 YBEY LSS FTCD SPATC1L PCBP3 PRMT2 LOC101928796 PCNT COL6A1 MCM3AP-AS1 |
nsv1065227 | Chr.21:46565742 - 46616312 on Build GRCh38 | Gain | DIP2A S100B |
nsv1057566 | Chr.21:46495670 - 46609786 on Build GRCh38 | Loss | DIP2A S100B |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |