Assay Details
Target Gene Details
Entrez Gene ID: | 9219 |
Gene Name: | metastasis associated 1 family member 2 |
Gene Aliases: |
MTA1L1, PID |
Location: |
Chr.11:62593203-62601840 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTA2 | NM_004739.3 | NP_004730.2 | ||
XM_017018561.1 | XP_016874050.1 | |||
AB016591.1 | BAA36707.1 | |||
AF295807.1 | AAG02241.1 | |||
AK301569.1 | ||||
BC053650.1 | AAH53650.1 | |||
CR749481.1 | CAH18309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain | AHNAK EML3 GANAB EEF1G TUT1 ROM1 B3GAT3 MTA2 MIR6747 |
nsv832180 | Chr.11:62434133 - 62597356 on Build GRCh38 | Loss | AHNAK EEF1G TUT1 MTA2 MIR6747 |
nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain | EML3 GANAB EEF1G TUT1 ROM1 B3GAT3 MTA2 MIR6747 |
nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss | TUT1 ROM1 BSCL2 B3GAT3 MTA2 MIR6747 AHNAK UQCC3 EML3 GANAB UBXN1 EEF1G SNORA57 LBHD1 METTL12 LRRN4CL C11orf98 HNRNPUL2-BSCL2 INTS5 |
nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain | TTC9C TUT1 ROM1 BSCL2 B3GAT3 HNRNPUL2 MTA2 MIR6747 UQCC3 EML3 GANAB UBXN1 EEF1G SNORA57 LBHD1 METTL12 LRRN4CL C11orf98 HNRNPUL2-BSCL2 GNG3 INTS5 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |