Assay Details
Target Gene Details
Entrez Gene ID: | 4664 |
Gene Name: | NGFI-A binding protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:190648860-190692766 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NAB1 | NM_001321312.1 | 3 | 1178 | NP_001308241.1 |
NM_001321313.1 | 3 | 964 | NP_001308242.1 | |
NM_001321314.1 | 2 | 887 | NP_001308243.1 | |
NM_001321315.1 | 2 | 887 | NP_001308244.1 | |
NM_005966.3 | 5 | 1440 | NP_005957.2 | |
XM_005246582.1 | 4 | 1113 | XP_005246639.1 | |
XM_005246583.1 | 4 | 1227 | XP_005246640.1 | |
XM_005246586.1 | 3 | 920 | XP_005246643.1 | |
XM_005246587.1 | 3 | 920 | XP_005246644.1 | |
XM_011511219.2 | 4 | 1082 | XP_011509521.1 | |
XM_017004170.1 | 4 | 1123 | XP_016859659.1 | |
XM_017004171.1 | 3 | 1293 | XP_016859660.1 | |
XM_017004172.1 | 3 | 1065 | XP_016859661.1 | |
XM_017004173.1 | 4 | 1148 | XP_016859662.1 | |
XM_017004174.1 | 3 | 964 | XP_016859663.1 | |
XM_017004175.1 | 3 | 1129 | XP_016859664.1 | |
XM_017004176.1 | 3 | 1032 | XP_016859665.1 | |
AF045451.1 | 2 | 887 | AAC25085.1 | |
AF045452.1 | 2 | 887 | AAC25086.1 | |
AK292446.1 | 3 | 978 | ||
AK296549.1 | 2 | 215 | ||
AK313542.1 | 2 | 985 | ||
BC035724.1 | 5 | 1329 | AAH35724.1 | |
U47007.1 | 2 | 887 | AAC50588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3584252 | Chr.2:190654887 - 190674560 on Build GRCh38 | Loss | NAB1 |
nsv3082 | Chr.2:190627513 - 190688068 on Build GRCh38 | Insertion | NAB1 |
nsv834492 | Chr.2:190574138 - 190781895 on Build GRCh38 | Gain | NAB1 NEMP2 |
dgv1200e212 | Chr.2:190638342 - 190674560 on Build GRCh38 | Loss | NAB1 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |