Assay Details
Target Gene Details
Entrez Gene ID: | 23398 |
Gene Name: | peptidylprolyl isomerase domain and WD repeat containing 1 |
Gene Aliases: |
- |
Location: |
Chr.5:65563236-65587549 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPWD1 | NM_001278926.1 | 7 | 1131 | NP_001265855.1 |
NM_001278927.1 | 6 | 921 | NP_001265856.1 | |
NM_001278929.1 | 5 | 991 | NP_001265858.1 | |
NM_015342.3 | 6 | 1097 | NP_056157.1 | |
XM_005248474.3 | 2 | 264 | XP_005248531.1 | |
XM_006714583.3 | 7 | 1067 | XP_006714646.1 | |
XM_011543295.1 | 5 | 1379 | XP_011541597.1 | |
XM_017009286.1 | 5 | 421 | XP_016864775.1 | |
AK025679.1 | 6 | 1044 | ||
AK298175.1 | 6 | 861 | ||
AK300022.1 | 7 | 1074 | ||
AK301654.1 | 5 | 931 | ||
BC015385.1 | 6 | 1025 | AAH15385.1 | |
D38552.1 | 6 | 1020 | BAA07555.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |