Assay Details
Target Gene Details
Entrez Gene ID: | 122809 |
Gene Name: | suppressor of cytokine signaling 4 |
Gene Aliases: |
SOCS7 |
Location: |
Chr.14:55027126-55049489 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SOCS4 | NM_080867.2 | 2 | 1442 | NP_543143.1 |
NM_199421.1 | 3 | 1571 | NP_955453.1 | |
XM_011536425.1 | 3 | 1625 | XP_011534727.1 | |
XM_011536426.1 | 3 | 1540 | XP_011534728.1 | |
AF424815.1 | 1 | 1222 | AAL60517.1 | |
AK124083.1 | 2 | 1295 | ||
AK127299.1 | 1 | 320 | ||
BC060790.1 | 3 | 1449 | AAH60790.1 | |
BX161418.1 | 2 | 1338 | CAD61893.1 | |
BX648734.1 | 2 | 1228 | ||
CR933659.1 | 2 | 1332 | CAI45958.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |