Assay Details
Target Gene Details
Entrez Gene ID: | 10766 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
APRO5, TOB4, TOBL, TROB2 |
Location: |
Chr.22:41433488-41448230 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOB2 | NM_016272.3 | 2 | 2093 | NP_057356.1 |
XM_005261315.2 | 2 | 3304 | XP_005261372.1 | |
XM_006724105.3 | 2 | 2610 | XP_006724168.1 | |
XM_017028539.1 | 2 | 4386 | XP_016884028.1 | |
AB051450.1 | 2 | 1852 | ||
BC038957.1 | 2 | 1866 | AAH38957.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834207 | Chr.22:41282650 - 41463790 on Build GRCh38 | Loss | ZC3H7B LOC100996598 TOB2 RANGAP1 PHF5A TEF |
nsv516925 | Chr.22:41252498 - 41585953 on Build GRCh38 | Loss | LOC105373044 POLR3H ZC3H7B LOC100996598 TOB2 PMM1 MIR6889 CSDC2 RANGAP1 PHF5A TEF ACO2 |
nsv834208 | Chr.22:41363612 - 41554028 on Build GRCh38 | Loss | LOC105373044 POLR3H LOC100996598 TOB2 PHF5A TEF ACO2 |
nsv589050 | Chr.22:41231539 - 41454496 on Build GRCh38 | Gain | ZC3H7B LOC100996598 TOB2 MIR6889 RANGAP1 CHADL TEF |
nsv524585 | Chr.22:40993505 - 41794305 on Build GRCh38 | Loss | LOC100996598 DESI1 PMM1 XRCC6 CHADL PHF5A L3MBTL2 LOC105373044 POLR3H EP300 ZC3H7B EP300-AS1 SNORD140 TOB2 MIR6889 MEI1 CSDC2 RANGAP1 SNU13 MIR1281 TEF C22orf46 ACO2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |