Assay Details
Target Gene Details
Entrez Gene ID: | 2621 |
Gene Name: | growth arrest specific 6 |
Gene Aliases: |
AXLLG, AXSF |
Location: |
Chr.13:113820549-113864103 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAS6 | NM_000820.3 | 14 | 1895 | NP_000811.1 |
AK092028.1 | 6 | 1867 | ||
AK122969.1 | 8 | 1532 | ||
AK126533.1 | 13 | 3179 | BAC86580.1 | |
AK290803.1 | 14 | 1784 | ||
AK303145.1 | 11 | 1441 | ||
AL833761.1 | 1 | 1067 | CAH56234.1 | |
BC038984.1 | 14 | 1843 | AAH38984.1 | |
BQ181504.1 | 2 | 657 | ||
L13720.1 | 14 | 1846 | AAA58494.1 |
Target Gene Details
Entrez Gene ID: | 650669 |
Gene Name: | GAS6 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:113815610-113842841 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GAS6-AS1 | NR_044995.2 | |||
BX385300.2 | ||||
DA460725.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv563390 | Chr.13:113809317 - 113845944 on Build GRCh38 | Loss | GAS6-AS1 GAS6 TMEM255B |
nsv1043348 | Chr.13:113756321 - 113867896 on Build GRCh38 | Gain | GAS6-AS1 GAS6 GAS6-AS2 LOC105377805 TMEM255B |
nsv951933 | Chr.13:113808028 - 113844427 on Build GRCh38 | Deletion | GAS6-AS1 GAS6 TMEM255B |
dgv3450n54 | Chr.13:113754200 - 113886735 on Build GRCh38 | Gain | LINC00454 GAS6-AS1 GAS6 GAS6-AS2 LOC105377805 TMEM255B |
esv24756 | Chr.13:113802553 - 113834906 on Build GRCh38 | Gain+Loss | GAS6-AS1 GAS6 TMEM255B |
nsv832727 | Chr.13:113753115 - 113918257 on Build GRCh38 | Loss | LINC00454 GAS6-AS1 LINC00452 GAS6 GAS6-AS2 LOC105377805 TMEM255B |
esv3582860 | Chr.13:113756381 - 113867871 on Build GRCh38 | Gain | GAS6-AS1 GAS6 GAS6-AS2 LOC105377805 TMEM255B |
nsv826836 | Chr.13:113753163 - 113875076 on Build GRCh38 | Loss | GAS6-AS1 GAS6 GAS6-AS2 LOC105377805 TMEM255B |
nsv563392 | Chr.13:113814175 - 113823135 on Build GRCh38 | Loss | GAS6-AS1 GAS6 |
nsv826837 | Chr.13:113802020 - 113834906 on Build GRCh38 | Loss | GAS6-AS1 GAS6 TMEM255B |
esv3633622 | Chr.13:113808937 - 113835264 on Build GRCh38 | Gain | GAS6-AS1 GAS6 TMEM255B |
esv3633615 | Chr.13:113724844 - 113843481 on Build GRCh38 | Gain | LINC00552 GAS6-AS1 GRK1 GAS6 LOC105377805 TMEM255B |
More Information
Set Membership: |
Intragenic Exonic Intronic Non-exonic DGV Variation |