Assay Details
Target Gene Details
Entrez Gene ID: | 7852 |
Gene Name: | C-X-C motif chemokine receptor 4 |
Gene Aliases: |
CD184, D2S201E, FB22, HM89, HSY3RR, LAP-3, LAP3, LCR1, LESTR, NPY3R, NPYR, NPYRL, NPYY3R, WHIM, WHIMS |
Location: |
Chr.2:136114349-136118155 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CXCR4 | NM_001008540.1 | 1 | 510 | NP_001008540.1 |
NM_003467.2 | 2 | 289 | NP_003458.1 | |
AF025375.1 | 1 | 194 | AAB81970.1 | |
AF147204.1 | 1 | 548 | AAF00130.1 | |
AF348491.1 | 2 | 237 | AAK29630.1 | |
AK129916.1 | 2 | 283 | ||
AK296674.1 | 2 | 283 | ||
AK309641.1 | 1 | 438 | ||
AK309885.1 | 2 | 388 | ||
AK312244.1 | 2 | 283 | ||
AY242129.1 | 1 | 194 | AAO92296.1 | |
BC020968.2 | 2 | 247 | AAH20968.1 | |
BT006660.1 | 1 | 194 | AAP35306.1 | |
D10924.1 | 2 | 270 | BAA01722.1 | |
KU245646.1 | 2 | 407 | ||
KU245647.1 | 2 | 293 | ||
L01639.1 | 2 | 219 | AAA16594.1 | |
L06797.1 | 2 | 266 | AAA03209.1 | |
M99293.1 | 2 | 278 | AAA16617.1 | |
X71635.1 | 2 | 261 | CAA50641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523997 | Chr.2:136110306 - 136463173 on Build GRCh38 | Gain | CXCR4 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |