Assay Details
Target Gene Details
Entrez Gene ID: | 64412 |
Gene Name: | GDNF inducible zinc finger protein 1 |
Gene Aliases: |
ZBTB23, ZNF336 |
Location: |
Chr.20:23361585-23373062 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GZF1 | NM_001317012.1 | 7 | 3025 | NP_001303941.1 |
NM_001317019.1 | 5 | 2776 | NP_001303948.1 | |
NM_022482.4 | 6 | 2928 | NP_071927.1 | |
XM_005260800.2 | 6 | 2882 | XP_005260857.1 | |
XM_011529321.2 | 6 | 2900 | XP_011527623.1 | |
XM_011529322.1 | 6 | 2861 | XP_011527624.1 | |
XM_017028017.1 | 6 | 2873 | XP_016883506.1 | |
XM_017028018.1 | 6 | 2815 | XP_016883507.1 | |
XM_017028019.1 | 5 | 2703 | XP_016883508.1 | |
AK025447.1 | 5 | 1845 | BAB15134.1 | |
AK056477.1 | 6 | 2878 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |