Assay Details
Target Gene Details
Entrez Gene ID: | 60343 |
Gene Name: | family with sequence similarity 3 member A |
Gene Aliases: |
2.19, DLD, DXS560S, XAP-7 |
Location: |
Chr.X:154506159-154516249 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss | TMLHE LOC101927830 FAM223A HCFC1 CTAG2 PLXNA3 GDI1 CTAG1A MPP1 GAB3 FAM50A MIR1184-2 MIR3202-1 H2AFB3 TEX28 LOC107987332 LOC105373386 TKTL1 H2AFB2 FLNA H2AFB1 SPRY3 VBP1 MIR3202-2 TAZ G6PD OPN1MW SMIM9 F8A1 FAM3A MIR1184-1 FAM223B CH17-340M24.3 RAB39B SLC10A3 F8A3 RPL10 MTCP1 CLIC2 UBL4A OPN1LW TMLHE-AS1 EMD MIR664B SNORA36A ATP6AP1 LAGE3 DKC1 BRCC3 MECP2 HCFC1-AS1 OPN1MW3 F8A2 CTAG1B MIR1184-3 F8 MIR6858 OPN1MW2 DNASE1L1 MIR718 TMEM187 FUNDC2 SNORA70 IKBKG SNORA56 CMC4 IRAK1 |
nsv519042 | Chr.X:154082978 - 154601444 on Build GRCh38 | Gain | FAM223A CH17-340M24.3 SLC10A3 PLXNA3 RPL10 GDI1 CTAG1A FAM50A UBL4A OPN1LW EMD ATP6AP1 LAGE3 TEX28 LOC107987332 LOC105373386 MECP2 OPN1MW3 TKTL1 MIR6858 FLNA OPN1MW2 DNASE1L1 SNORA70 TAZ IKBKG G6PD OPN1MW FAM3A |
esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss | PLXNB3 FAM223A PNMA6A HCFC1 PLXNA3 GDI1 CTAG1A ABCD1 MAGEA1 FAM50A MIR3202-1 ARHGAP4 PNCK AVPR2 TEX28 LOC107987332 LOC105373386 LOC105373377 TKTL1 FLNA MIR3202-2 TAZ BCAP31 L1CAM G6PD OPN1MW LOC105373381 LOC105373383 FAM3A SRPK3 IDH3G ATP2B3 CH17-340M24.3 SLC10A3 LCA10 FAM58A RPL10 TREX2 LOC107983954 ZNF275 UBL4A OPN1LW DUSP9 BGN EMD HAUS7 ATP6AP1 LAGE3 PDZD4 MECP2 HCFC1-AS1 ZFP92 OPN1MW3 SLC6A8 LOC105373378 MIR6858 OPN1MW2 NAA10 DNASE1L1 MIR718 SSR4 TMEM187 SNORA70 IKBKG RENBP IRAK1 |
nsv508816 | Chr.X:154492171 - 154591783 on Build GRCh38 | Insertion | FAM223A IKBKG G6PD FAM3A CTAG1A |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |