Assay Details
Target Gene Details
Entrez Gene ID: | 84134 |
Gene Name: | translocase of outer mitochondrial membrane 40 like |
Gene Aliases: |
TOMM40B |
Location: |
Chr.1:161225939-161230746 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TOMM40L | NM_001286373.1 | 9 | 1230 | NP_001273302.1 |
NM_001286374.1 | 8 | 1137 | NP_001273303.1 | |
NM_032174.5 | 10 | 1332 | NP_115550.2 | |
XM_006711572.2 | 10 | 1207 | XP_006711635.1 | |
XM_011510057.2 | 8 | 900 | XP_011508359.1 | |
XM_017002481.1 | 10 | 1170 | XP_016857970.1 | |
XM_017002482.1 | 9 | 1098 | XP_016857971.1 | |
AK022832.1 | 7 | 948 | BAB14265.1 | |
AK027704.1 | 10 | 1211 | BAB55309.1 | |
AK297831.1 | 9 | 984 | ||
AK297848.1 | 9 | 1109 | ||
AL523459.3 | ||||
BC014946.1 | 9 | 1185 | AAH14946.1 | |
BX537951.1 | 10 | 1209 | CAD97918.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |