Assay Details
Target Gene Details
Entrez Gene ID: | 2217 |
Gene Name: | Fc fragment of IgG receptor and transporter |
Gene Aliases: |
FCRN, alpha-chain |
Location: |
Chr.19:49512279-49526428 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FCGRT | NM_001136019.2 | 3 | 707 | NP_001129491.1 |
NM_004107.4 | 2 | 443 | NP_004098.1 | |
XM_005258657.1 | 2 | 262 | XP_005258714.1 | |
XM_011526627.2 | 2 | 400 | XP_011524929.1 | |
AK074734.1 | 1 | 835 | ||
AK075510.1 | 2 | 254 | ||
AK075532.1 | 3 | 357 | ||
AK299124.1 | 2 | 227 | ||
BC008734.2 | 2 | 387 | AAH08734.1 | |
BI520292.1 | 3 | 706 | ||
BT006828.1 | 2 | 221 | AAP35474.1 | |
BX647163.1 | 3 | 327 | CAI46032.1 | |
DC312637.1 | 2 | 444 | ||
U12255.1 | 2 | 346 | AAA58958.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | SNAR-G2 CCDC155 FLJ26850 SNAR-C3 MIR6798 NUCB1-AS1 RPL18 PIH1D1 ELSPBP1 PLA2G4C-AS1 SNAR-A11 RASIP1 SNAR-A7 SNAR-C4 CGB5 SNORD35A TBC1D17 PRR12 SLC17A7 SULT2B1 GYS1 BSPH1 ADM5 IL4I1 SNAR-D SNAR-G1 HSD17B14 SNAR-A14 LMTK3 SCAF1 ZNF114 CGB8 LIN7B C19orf68 PLEKHA4 CYTH2 FAM83E SPACA4 CA11 AP2A1 DKKL1 NUCB1 BCAT2 LOC105447645 SNAR-A9 SNAR-C5 SNAR-A10 BCL2L12 LOC105372430 SPHK2 GRIN2D LOC101928295 SNAR-A1 EMP3 NTF4 PRMT1 TSKS RUVBL2 SIGLEC16 TMEM143 PPFIA3 TULP2 CGB2 PTOV1-AS2 SNORD35B SNORD34 MIR5088 FUT1 ATF5 DHDH ALDH16A1 MIR4324 SNAR-C2 FUT2 FTL RCN3 C19orf73 CABP5 SIGLEC11 VRK3 SNAR-C1 DBP SNAR-A13 SNORD33 SYNGR4 SNAR-A2 SNAR-B2 SEC1P PNKP KCNA7 CARD8 ZNF473 RPS11 MIR4750 GRWD1 FCGRT MIR6799 CGB1 MED25 MIR4749 SNRNP70 PRRG2 SNAR-A8 SNAR-A3 CD37 MIR150 CCDC114 KCNJ14 RRAS LIG1 SNAR-A4 IZUMO1 NUP62 SNAR-A5 MIR4751 CGB3 MAMSTR MIR6800 NTN5 GFY LOC100287477 SNORD32A IRF3 HRC CARD8-AS1 FGF21 CGB7 PPP1R15A KDELR1 BAX SNAR-B1 TRPM4 AKT1S1 TEAD2 PLA2G4C SLC6A16 PTOV1 RPL13A PTH2 FLT3LG SNAR-A6 LOC101059948 PTOV1-AS1 LHB NOSIP CPT1C SNAR-A12 FUZ |
nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss | SNORD32A PRRG2 FLT3LG RCN3 MIR150 RPS11 FCGRT SNORD35B SNORD34 NOSIP SNORD35A SNORD33 PRR12 RPL13A |
nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss | SNORD32A FLT3LG RCN3 MIR150 RPS11 PIH1D1 FCGRT SNORD35B SNORD34 NOSIP SNORD35A SNORD33 RPL13A ALDH16A1 |
nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion | SNORD32A BCL2L12 IRF3 PRMT1 RPS11 TSKS FCGRT MIR6799 SNORD35B SNORD34 MIR5088 SNORD35A PRR12 MED25 ADM5 RPL13A PRRG2 RCN3 MIR150 SCAF1 PTOV1-AS1 RRAS AP2A1 NOSIP SNORD33 CPT1C MIR6800 FUZ |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |