Assay Details
Target Gene Details
Entrez Gene ID: | 80380 |
Gene Name: | programmed cell death 1 ligand 2 |
Gene Aliases: |
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2 |
Location: |
Chr.9:5510438-5571282 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PDCD1LG2 | NM_025239.3 | 3 | 559 | NP_079515.2 |
XM_005251600.3 | 3 | 666 | XP_005251657.1 | |
AF329193.1 | 3 | 544 | AAK31105.1 | |
AF344424.1 | 3 | 559 | AAK15370.1 | |
AK001872.1 | ||||
AY254343.1 | 2 | 286 | AAP13471.1 | |
AY271901.1 | 2 | 286 | AAP49000.1 | |
AY271902.1 | 2 | 286 | AAP49001.1 | |
BC074766.2 | 3 | 320 | AAH74766.1 | |
BC113678.1 | 3 | 322 | ||
BC113680.1 | 3 | 322 | ||
BX951459.1 | 1 | 196 | ||
DQ336699.1 | 2 | 253 | ABC59616.1 | |
EF444806.1 | 3 | 406 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509292 | Chr.9:5469291 - 5546768 on Build GRCh38 | Insertion | PDCD1LG2 CD274 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |