Assay Details
Target Gene Details
Entrez Gene ID: | 6725 |
Gene Name: | src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites |
Gene Aliases: |
C20orf148, PTK70, SRM, dJ697K14.1 |
Location: |
Chr.20:63539924-63547504 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SRMS | NM_080823.3 | NP_543013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv953306 | Chr.20:63151949 - 63571247 on Build GRCh38 | Deletion | HELZ2 LOC107985425 MIR124-3 SRMS PTK6 BIRC7 FNDC11 KCNQ2 MIR4326 COL20A1 CHRNA4 MIR3196 FLJ16779 LOC102723788 LOC100130152 EEF1A2 NKAIN4 PPDPF ARFGAP1 YTHDF1 LOC100130587 |
nsv586573 | Chr.20:63535645 - 63560584 on Build GRCh38 | Loss | HELZ2 SRMS PTK6 FNDC11 |
nsv521734 | Chr.20:63522433 - 63566995 on Build GRCh38 | Loss | HELZ2 SRMS PTK6 FNDC11 |
nsv9825 | Chr.20:62775470 - 63601330 on Build GRCh38 | Gain | LOC107985428 HAR1B GID8 SLC17A9 PTK6 OGFR-AS1 DPH3P1 BIRC7 FNDC11 TCFL5 MIR3196 LOC102723788 LINC00029 COL9A3 LOC63930 HELZ2 HAR1A LOC107985425 MIR124-3 SRMS LINC01056 KCNQ2 MIR4326 COL20A1 CHRNA4 MRGBP DIDO1 FLJ16779 LINC00659 LOC100130152 EEF1A2 OGFR NKAIN4 BHLHE23 PPDPF ARFGAP1 YTHDF1 GMEB2 LOC100130587 |
nsv470563 | Chr.20:63486715 - 63793308 on Build GRCh38 | Loss | ZBTB46 HELZ2 SRMS RTEL1-TNFRSF6B PTK6 FNDC11 LOC100505771 ARFRP1 ZGPAT STMN3 TNFRSF6B SLC2A4RG LIME1 EEF1A2 PPDPF RTEL1 GMEB2 |
nsv834033 | Chr.20:63421436 - 63554987 on Build GRCh38 | Gain | SRMS EEF1A2 PTK6 PPDPF FNDC11 KCNQ2 |
nsv834034 | Chr.20:63462825 - 63625901 on Build GRCh38 | Loss | HELZ2 SRMS EEF1A2 PTK6 PPDPF FNDC11 KCNQ2 GMEB2 |
nsv586574 | Chr.20:63540866 - 63547263 on Build GRCh38 | Loss | SRMS |
nsv586569 | Chr.20:63496168 - 63548764 on Build GRCh38 | Loss | SRMS EEF1A2 PTK6 PPDPF |
nsv517702 | Chr.20:63540916 - 63581575 on Build GRCh38 | Loss | HELZ2 SRMS FNDC11 |
esv2758806 | Chr.20:63470996 - 63745014 on Build GRCh38 | Gain+Loss | ZBTB46 HELZ2 SRMS RTEL1-TNFRSF6B PTK6 FNDC11 KCNQ2 LOC100505771 ARFRP1 ZGPAT STMN3 TNFRSF6B SLC2A4RG LIME1 EEF1A2 PPDPF RTEL1 GMEB2 |
More Information
Set Membership: |
Intragenic Non-exonic DGV Variation |