Assay Details
Target Gene Details
Entrez Gene ID: | 84167 |
Gene Name: | chromosome 19 open reading frame 44 |
Gene Aliases: |
- |
Location: |
Chr.19:16496311-16521369 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
C19orf44 | NM_032207.3 | 9 | 2533 | NP_115583.1 |
XM_006722920.3 | 8 | 2437 | XP_006722983.1 | |
AK025395.1 | 9 | 2431 | BAB15125.1 | |
BC027869.1 | 9 | 2352 | AAH27869.1 |
Target Gene Details
Entrez Gene ID: | 10523 |
Gene Name: | calcium homeostasis endoplasmic reticulum protein |
Gene Aliases: |
DAN16, SCAF6, SRA1 |
Location: |
Chr.19:16517889-16542452 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHERP | NM_006387.5 | 14 | 2327 | NP_006378.3 |
AB209730.1 | 5 | 704 | BAD92967.1 | |
AF536542.1 | 14 | 2294 | AAN77183.1 | |
AK296883.1 | 8 | 1038 | ||
AK298408.1 | 6 | 1055 | ||
BC021294.2 | 14 | 2312 | AAH21294.1 | |
U94836.2 | 13 | 2229 | AAB53327.1 |
More Information
Set Membership: |
Intragenic Exonic non-DGV Variation |