Assay Details
Target Gene Details
Entrez Gene ID: | 27158 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
NR1, bA350O14.9 |
Location: |
Chr.9:137205667-137219361 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDOR1 | NM_001144026.2 | 14 | 2536 | NP_001137498.1 |
NM_001144027.2 | 13 | 2385 | NP_001137499.1 | |
NM_001144028.2 | 14 | 2488 | NP_001137500.1 | |
NM_014434.3 | 14 | 2509 | NP_055249.1 | |
XM_011518545.1 | 13 | 2424 | XP_011516847.1 | |
XM_011518547.1 | 14 | 2326 | XP_011516849.1 | |
XM_011518548.2 | 13 | 3726 | XP_011516850.1 | |
XM_017014627.1 | 14 | 2487 | XP_016870116.1 | |
AF199509.1 | AAF25205.1 | |||
AK026089.1 | ||||
AK074403.1 | ||||
AK225995.1 | ||||
BC015735.1 | AAH15735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv13010n54 | Chr.9:137191539 - 137233413 on Build GRCh38 | Loss | SLC34A3 RNF208 TPRN CYSRT1 NDOR1 TMEM203 RNF224 |
nsv616012 | Chr.9:137197680 - 137223834 on Build GRCh38 | Loss | RNF208 TPRN NDOR1 TMEM203 |
nsv616007 | Chr.9:137079368 - 137240666 on Build GRCh38 | Loss | SLC34A3 MAN1B1-AS1 MIR3621 RNF208 UAP1L1 CYSRT1 SSNA1 ANAPC2 TMEM210 RNF224 LOC101930307 LRRC26 TPRN MAN1B1 GRIN1 NDOR1 TMEM203 DPP7 |
dgv13009n54 | Chr.9:137173890 - 137327859 on Build GRCh38 | Loss | SLC34A3 STPG3 FAM166A NRARP RNF208 CYSRT1 TUBB4B SSNA1 ANAPC2 TOR4A NELFB RNF224 EXD3 TPRN NDOR1 C9orf173-AS1 TMEM203 |
nsv524322 | Chr.9:136795692 - 137482216 on Build GRCh38 | Loss | STPG3 NRARP UAP1L1 MIR4292 FUT7 PHPT1 CCDC183-AS1 MIR7114 GRIN1 C9orf173-AS1 FBXW5 EDF1 SAPCD2 MAN1B1-AS1 MIR4479 C9orf142 LCN12 C8G SSNA1 ANAPC2 TOR4A TMEM210 EXD3 TPRN C9orf139 NDOR1 ABCA2 NSMF FAM166A MIR3621 RNF208 PRR31 CYSRT1 TUBB4B NELFB RNF224 RABL6 CLIC3 C9orf172 LOC101930307 LRRC26 ENTPD8 PNPLA7 ENTPD2 NPDC1 SLC34A3 LCNL1 NOXA1 PTGDS MAN1B1 MAMDC4 TMEM203 DPP7 CCDC183 TRAF2 |
nsv951207 | Chr.9:137136949 - 137379148 on Build GRCh38 | Deletion | SLC34A3 STPG3 FAM166A MIR3621 NRARP RNF208 CYSRT1 TUBB4B SSNA1 ANAPC2 TOR4A TMEM210 NELFB RNF224 LRRC26 EXD3 TPRN GRIN1 NDOR1 C9orf173-AS1 TMEM203 |
esv2739415 | Chr.9:137216879 - 137217380 on Build GRCh38 | Deletion | NDOR1 |
nsv1161918 | Chr.9:137118319 - 137438026 on Build GRCh38 | Duplication | SLC34A3 STPG3 FAM166A MIR3621 NRARP RNF208 NOXA1 CYSRT1 TUBB4B SSNA1 ANAPC2 TOR4A TMEM210 NELFB RNF224 LRRC26 EXD3 TPRN GRIN1 ENTPD8 NDOR1 C9orf173-AS1 TMEM203 |
esv2739416 | Chr.9:137216942 - 137217090 on Build GRCh38 | Deletion | NDOR1 |
esv2422220 | Chr.9:136584135 - 137350098 on Build GRCh38 | Deletion | STPG3 NRARP FAM69B UAP1L1 MIR4292 FUT7 PHPT1 EGFL7 CCDC183-AS1 GRIN1 C9orf173-AS1 FBXW5 LCN8 EDF1 SAPCD2 MAN1B1-AS1 SNHG7 MIR4479 C9orf142 LCN12 C8G SSNA1 ANAPC2 MIR126 TOR4A TMEM210 AGPAT2 LINC01451 EXD3 TPRN C9orf139 TMEM141 NDOR1 ABCA2 LCN6 LCN10 FAM166A MIR3621 RNF208 MIR6722 PRR31 CYSRT1 TUBB4B NELFB RNF224 RABL6 CLIC3 C9orf172 LOC101930307 LRRC26 HSPC324 SNORA17B ENTPD2 NPDC1 SLC34A3 LCNL1 LCN15 LOC100128593 PTGDS MAN1B1 MAMDC4 TMEM203 DPP7 CCDC183 SNORA17A TRAF2 |
More Information
Set Membership: |
Intragenic Exonic DGV Variation |